EMAIL THIS PAGE TO A FRIEND
288713 Sigma-Aldrich

Florisil®

TLC grade, ~2 μm avg. part. size

Synonym: Activated magnesium silicate, Magnesium silicate

Purchase

Description

Legal Information

Florisil is a registered trademark of U.S. Silica Company

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?