• USA Home
  • 28945 - Cyclohexanecarboxylic acid

EMAIL THIS PAGE TO A FRIEND
28945 Sigma-Aldrich

Cyclohexanecarboxylic acid

purum, ≥97.0% (GC)

Synonym: Hexahydrobenzoic acid

  • CAS Number 98-89-5

  • Linear Formula C6H11CO2H

  • Molecular Weight 128.17

  •  Beilstein Registry Number 970529

  •  EC Number 202-711-3

  •  MDL number MFCD00001461

  •  PubChem Substance ID 329753297

Purchase

Properties

Related Categories Building Blocks, C7, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   NZNMSOFKMUBTKW-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.461(lit.)
bp   232-233 °C(lit.)
mp   29-31 °C(lit.)
density   1.033 g/mL at 25 °C(lit.)

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
GU8370000
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?