• USA Home
  • 28980 - Dimethyl cyclohexane-1,4-dicarboxylate

EMAIL THIS PAGE TO A FRIEND
28980 Sigma-Aldrich

Dimethyl cyclohexane-1,4-dicarboxylate

technical, cis+trans, ≥90% (GC)

  • CAS Number 94-60-0

  • Empirical Formula (Hill Notation) C10H16O4

  • Molecular Weight 200.23

  •  Beilstein Registry Number 1876703

  •  EC Number 202-347-5

  •  MDL number MFCD00001460

  •  PubChem Substance ID 329753301

Purchase

Properties

Related Categories Building Blocks, C10 to C11, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   LNGAGQAGYITKCW-UHFFFAOYSA-N
grade   technical
assay   ≥90% (GC)
refractive index   n20/D 1.458
bp   132 °C/10 mmHg(lit.)
density   1.105 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

206431 Dimethyl cyclohexane-1,4-dicarboxylate, mixture of cis and trans, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?