289825 Sigma-Aldrich

Aluminum acetate, basic

DOWNLOAD MSDS (PDF)

Synonym: Aluminum diacetate hydroxide, Aluminum hydroxide acetate

  • CAS Number 142-03-0

  • Linear Formula Al(OH)(C2H3O2)2

  • Molecular Weight 162.08

  •  EC Number 205-518-2

  •  MDL number MFCD00008688

  •  PubChem Substance ID 24857215

Description

Other Notes

No stabilizers added.

Packaging

100, 500 g in glass bottle

Physical form

Insoluble form

Price and Availability

Customers Also Viewed

Aluminum acetate, dibasic

contains boric acid as stabilizer

Aluminum acetylacetonate

ReagentPlus®, 99%

Zinc acetate

99.99% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?