• USA Home
  • 29002 - (1R,2R)-trans-1,2-Cyclohexanediol

EMAIL THIS PAGE TO A FRIEND
29002 Sigma-Aldrich

(1R,2R)-trans-1,2-Cyclohexanediol

puriss., ≥99.0% (sum of enantiomers, GC)

Synonym: (1R)-trans-1,2-Cyclohexanediol

  • CAS Number 1072-86-2

  • Linear Formula C6H10(OH)2

  • Molecular Weight 116.16

  •  Beilstein Registry Number 4780913

  •  MDL number MFCD00066430

  •  PubChem Substance ID 329753306

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Polyols More...
InChI Key   PFURGBBHAOXLIO-PHDIDXHHSA-N
grade   puriss.
assay   ≥99.0% (sum of enantiomers, GC)
optical activity   [α]20/D −39±1°, c = 1.6% in H2O
optical purity   enantiomeric ratio: ≥99:1 (GC)
mp   106-109 °C
  107-109 °C(lit.)

Description

Other Notes

Chiral auxiliary for conjugate addition reactions; Asymmetric alkylation via chiral acetals

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
273.2 °F
Flash Point(C) 
134 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of trans-1,2-Cyclohexanediol Enantiomers on Astec® CHIRALDEX® B-PH

From our library of Articles, Sigma-Aldrich presents GC Analysis of trans-1,2-Cyclohexanediol Enantiomers on Astec® CHIRALDEX® B-PH
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

421790 (1R,2R)-trans-1,2-Cyclohexanediol, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?