EMAIL THIS PAGE TO A FRIEND
29070 Sigma-Aldrich

1,4-Cyclohexanedione

purum, ≥98.0% (GC)

Purchase

Properties

Related Categories Building Blocks, C3 to C6, Carbonyl Compounds, Chemical Synthesis, Ketones,
InChI Key   DCZFGQYXRKMVFG-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
mp   76-79 °C
  77-78.5 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
269.6 °F
Flash Point(C) 
132 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

125423 1,4-Cyclohexanedione, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?