• USA Home
  • 29224 - 4-(Dimethylamino)pyridine

EMAIL THIS PAGE TO A FRIEND
29224 Sigma-Aldrich

4-(Dimethylamino)pyridine

puriss., ≥99.0% (NT)

Synonym: N,N-Dimethylpyridin-4-amine, DMAP

  • CAS Number 1122-58-3

  • Empirical Formula (Hill Notation) C7H10N2

  • Molecular Weight 122.17

  •  Beilstein Registry Number 110354

  •  EC Number 214-353-5

  •  MDL number MFCD00006418

Purchase

Properties

Related Categories Acids & Bases, Bases, Chemical Synthesis, Organic Bases, Synthetic Reagents More...
InChI Key   VHYFNPMBLIVWCW-UHFFFAOYSA-N
grade   puriss.
assay   ≥99.0% (NT)
mp   108-110 °C(lit.)
  111-114 °C
solubility   methanol: soluble0.1 g/mL, clear

Description

Application

A highly efficient catalyst for acylation reactions

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
US9230000
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

Coupling Reagents

In principle, the seemingly simple formation of a peptide bond can be accomplished using all the procedures available in organic chemistry for the synthesis of carboxylic acid amides. However, due to...
Matthias Junkers
Aldrich ChemFiles 2007, 7.2, 3.
Keywords: Acylations, Catalysis, Condensations, Dehydration reaction, Peptide synthesis, Racemizations, Solid phase peptide synthesis

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?