EMAIL THIS PAGE TO A FRIEND
29236 Sigma-Aldrich

L-Alanine

purum, ≥98.0% (NT)

Synonym: (S)-2-Aminopropionic acid, L-α-Aminopropionic acid

  • CAS Number 56-41-7

  • Empirical Formula (Hill Notation) C3H7NO2

  • Molecular Weight 89.09

  •  Beilstein Registry Number 1720248

  •  EC Number 200-273-8

  •  MDL number MFCD00064410

  •  PubChem Substance ID 329753325

Purchase

Properties

Related Categories Alanine, Amino Acids & Derivatives, Chemical Biology, Chemical Synthesis, Peptide Chemistry More...
InChI Key   QNAYBMKLOCPYGJ-REOHCLBHSA-N
grade   purum
assay   ≥98.0% (NT)
optical activity   [α]20/D +13.5±1°, c = 5% in 5 M HCl
loss   ≤0.5% loss on drying

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?