EMAIL THIS PAGE TO A FRIEND
29244 Sigma-Aldrich

Anion HPCE-Kit

for HPCE

Synonym: Multi anion HPCE-Kit

Purchase

Properties

Related Categories Analytical Standards, Analytical/Chromatography, Anion Standards, Electrophoresis, Elemental, Ions and Water (Karl Fischer) Standards,
grade   for HPCE
shelf life   limited shelf life, expiry date on the label
format   multi-component solution

Description

Components

Composition:
82619 buffer solution pH 7.7 for HPCE, 100 mL
95283 water for HPCE, 10 mL
29235 multielement anion HPCE standard solution, 10 mL

Application

The buffer solution pH 7.7 for HPCE is designed for the analysis of anions by indirect UV detection with reversed electro-osmotic flow. The components can also be ordered separately under the product numbers indicated.

Other Notes

Determination of anions with capillary electrophoresis and indirect UV detection

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?