• USA Home
  • 29287 - (S)-(+)-1-Cyclohexylethylamine

EMAIL THIS PAGE TO A FRIEND
29287 Sigma-Aldrich

(S)-(+)-1-Cyclohexylethylamine

purum, ≥98.0% (sum of enantiomers, GC)

Synonym: (S)-(+)-α-Methylcyclohexanemethylamine

  • CAS Number 17430-98-7

  • Linear Formula C6H11CH(CH3)NH2

  • Molecular Weight 127.23

  •  Beilstein Registry Number 2935069

  •  MDL number MFCD00066261

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Derivatization Reagents, Derivatization Reagents HPLC, UV-VIS More...
InChI Key   XBWOPGDJMAJJDG-ZETCQYMHSA-N
grade   purum
assay   ≥98.0% (sum of enantiomers, GC)
optical activity   [α]20/D +3.8±0.3°, neat
refractive index   n20/D 1.4614(lit.)
  n20/D 1.463
bp   60 °C/12 mmHg(lit.)
density   0.856 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2734PSN1 3(8) / PGII
WGK Germany 
3
Flash Point(F) 
125.6 °F
Flash Point(C) 
52 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

336513 (S)-(+)-1-Cyclohexylethylamine, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?