293253 Sigma-Aldrich

Hexane, mixture of isomers

CHROMASOLV®, for HPLC, ≥98.5%

DOWNLOAD MSDS (PDF)

Synonym: Hexanes, Isohexane

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, CHROMASOLV for HPLC, Chromatography Reagents & Solvents, HPLC & UHPLC Solvents (CHROMASOLV),
grade   CHROMASOLV®
  for HPLC
vapor density   3 (vs air)
description   Mixture of hexane isomers plus methylcyclopentane
assay   ≥98.5%
autoignition temp.   437 °F
expl. lim.   7.5 %
impurities   ≤0.0003 meq/g water-soluble titr. acid
  ≤0.005% S compounds
  ≤0.02% water
evapn. residue   <0.0005%
color   APHA: ≤10
refractive index   n20/D 1.379(lit.)
bp   68-70 °C(lit.)
mp   −95 °C(lit.)
density   0.672 g/mL at 25 °C(lit.)
λ   H2O reference
UV absorption   λ: 195 nm Amax: 1.0
  λ: 210 nm Amax: 0.30
  λ: 220 nm Amax: 0.10
  λ: 230 nm Amax: 0.05
  λ: 240 nm Amax: 0.02
  λ: 250-400 nm Amax: 0.01
suitability   passes test for HPLC

Description

Application

Meets ACS specifications

Other Notes

Contains thiophene

Packaging

1 pkg in Pure-Pac™ 2

200 L in Pure-Pac™ 1

1, 6×1, 2, 4×2, 4, 4×4 L in glass bottle

50 L in Pure-Pac™ 2

View returnable container options.

Legal Information

CHROMASOLV is a registered trademark of Sigma-Aldrich Laborchemikalien GmbH

Price and Availability

Customers Also Viewed

Dichloromethane

CHROMASOLV®, for HPLC, ≥99.8%, contains amylene as stabilizer

Hexane, mixture of isomers

anhydrous, ≥99%

Hexane, mixture of isomers

CHROMASOLV® Plus, for HPLC, ≥98.5%

Hexane, mixture of isomers

CHROMASOLV® Plus, for HPLC, ≥98.5%

Hexane, mixture of isomers

CHROMASOLV®, for HPLC, ≥98.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?