• USA Home
  • 29352 - Barium cyclohexanebutyrate

EMAIL THIS PAGE TO A FRIEND
29352 Sigma-Aldrich

Barium cyclohexanebutyrate

oil soluble standard for atom spectroscopy

Synonym: Barium 4-cyclohexylbutyrate, Bis(cyclohexanebutanoic acid)barium salt, Cyclohexanebutyric acid barium salt

Purchase

Properties

grade   oil soluble standard for atom spectroscopy
InChI Key   NFAVTTVOXXCARJ-UHFFFAOYSA-L
form   neat
application(s)   ICP/AAS: suitable
mp   225 °C (dec.)(lit.)
format   neat

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1564 6.1/PG 3
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

228109 Barium cyclohexanebutyrate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?