• USA Home
  • 29459 - Cyclohexylmagnesium chloride solution

EMAIL THIS PAGE TO A FRIEND
29459 Sigma-Aldrich

Cyclohexylmagnesium chloride solution

technical, ~1.6 M in diethyl ether

  • CAS Number 931-51-1

  • Empirical Formula (Hill Notation) C6H11ClMg

  • Molecular Weight 142.91

  •  Beilstein Registry Number 3535440

  •  MDL number MFCD00003816

  •  PubChem Substance ID 329753336

Purchase

Properties

Related Categories Alkyl, Chemical Synthesis, Grignard Reagents, Organometallic Reagents
InChI Key   WMJMABVHDMRMJA-UHFFFAOYSA-M
grade   technical
concentration   ~1.6 M in diethyl ether
density   0.83 g/mL at 20 °C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 2924 3/PG 1
WGK Germany 
1
Flash Point(F) 
-40 °F
Flash Point(C) 
-40 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

224413 Cyclohexylmagnesium chloride solution, 2.0 M in diethyl ether

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?