EMAIL THIS PAGE TO A FRIEND
29600 Sigma-Aldrich

Cyclooctane

purum, ≥99.0% (GC)

  • CAS Number 292-64-8

  • Empirical Formula (Hill Notation) C8H16

  • Molecular Weight 112.21

  •  Beilstein Registry Number 1900349

  •  EC Number 206-031-8

  •  MDL number MFCD00004162

  •  PubChem Substance ID 329753380

Purchase

Properties

Related Categories Alkanes, Building Blocks, Chemical Synthesis, Cyclic, Organic Building Blocks More...
vapor pressure   16 mmHg ( 37.7 °C)
InChI Key   WJTCGQSWYFHTAC-UHFFFAOYSA-N
grade   purum
assay   ≥99.0% (GC)
refractive index   n20/D 1.458(lit.)
  n20/D 1.458
bp   151 °C/740 mmHg(lit.)
mp   10-13 °C(lit.)
density   0.834 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3295C 3 / PGIII
WGK Germany 
2
Flash Point(F) 
82.4 °F
Flash Point(C) 
28 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C109401 Cyclooctane, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?