EMAIL THIS PAGE TO A FRIEND
29660 Sigma-Aldrich

Cyclooctylamine

purum, ≥98.0% (GC)

Synonym: Aminocyclooctane

  • CAS Number 5452-37-9

  • Linear Formula C8H15NH2

  • Molecular Weight 127.23

  •  Beilstein Registry Number 2689120

  •  EC Number 226-694-7

  •  MDL number MFCD00001748

Purchase

Properties

InChI Key   HSOHBWMXECKEKV-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.482(lit.)
  n20/D 1.482
bp   190 °C(lit.)
mp   −48 °C(lit.)
density   0.928 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2735PSN1 8 / PGII
WGK Germany 
3
Flash Point(F) 
145.4 °F
Flash Point(C) 
63 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

C110604 Cyclooctylamine, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?