EMAIL THIS PAGE TO A FRIEND
29686 Sigma-Aldrich

Cyclopentane

B&J Brand (product of Burdick & Jackson)

  • CAS Number 287-92-3

  • Empirical Formula (Hill Notation) C5H10

  • Molecular Weight 70.13

  •  Beilstein Registry Number 1900195

  •  MDL number MFCD00001356

  •  PubChem Substance ID 329753386

Purchase

Properties

grade   B&J Brand (product of Burdick & Jackson)
vapor density   ~2 (vs air)
InChI Key   RGSFGYAAUTVSQA-UHFFFAOYSA-N
autoignition temp.   682 °F
expl. lim.   8.7 %
refractive index   n20/D 1.405(lit.)
bp   50 °C(lit.)
mp   −94 °C(lit.)
density   0.751 g/mL at 25 °C(lit.)

Description

Other Notes

Sales restrictions may apply

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?