• USA Home
  • 29723 - (1S,2S)-trans-1,2-Cyclopentanediol

EMAIL THIS PAGE TO A FRIEND
29723 Sigma-Aldrich

(1S,2S)-trans-1,2-Cyclopentanediol

purum, ≥98.0% (sum of enantiomers, GC)

Synonym: (1S)-trans-1,2-Cyclopentanediol

  • CAS Number 63261-45-0

  • Empirical Formula (Hill Notation) C5H10O2

  • Molecular Weight 102.13

  •  Beilstein Registry Number 2496900

  •  MDL number MFCD00082581

  •  PubChem Substance ID 329753387

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks, Polyols More...
InChI Key   VCVOSERVUCJNPR-WHFBIAKZSA-N
grade   purum
assay   ≥98.0% (sum of enantiomers, GC)
optical activity   [α]20/D +21±2°, c = 1% in chloroform
optical purity   enantiomeric ratio: ≥99.5:0.5 (GC)
mp   46-50 °C
storage temp.   2-8°C

Description

Other Notes

Diol used as chiral building block or as auxiliary

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?