EMAIL THIS PAGE TO A FRIEND
29770 Sigma-Aldrich

Cyclopentanone

ReagentPlus®, ≥99.0%

Purchase

Properties

Related Categories Building Blocks, C3 to C6, Carbonyl Compounds, Chemical Synthesis, Ketones,
grade   ReagentPlus®
InChI Key   BGTOWKSIORTVQH-UHFFFAOYSA-N
assay   ≥99.0%
impurities   ≤0.5% water
refractive index   n20/D 1.437(lit.)
  n20/D 1.437
bp   130-131 °C(lit.)
mp   −51 °C(lit.)
density   0.951 g/mL at 25 °C(lit.)

Description

Legal Information

ReagentPlus is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2245 3 / PGIII
WGK Germany 
1
RTECS 
GY4725000
Flash Point(F) 
86 °F
Flash Point(C) 
30 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?