EMAIL THIS PAGE TO A FRIEND
29826 Sigma-Aldrich

Cyclopentene oxide

purum, ≥98.0% (GC)

Synonym: 1,2-Epoxycyclopentane, 6-Oxabicyclo[3.1.0]hexane

  • CAS Number 285-67-6

  • Empirical Formula (Hill Notation) C5H8O

  • Molecular Weight 84.12

  •  Beilstein Registry Number 102495

  •  EC Number 206-005-6

  •  MDL number MFCD00005161

  •  PubChem Substance ID 329753392

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Epoxides, Organic Building Blocks, Oxygen Compounds More...
InChI Key   GJEZBVHHZQAEDB-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.434(lit.)
  n20/D 1.434
bp   102 °C(lit.)
density   0.964 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993BF 3 / PGII
WGK Germany 
3
RTECS 
RN8935000
Flash Point(F) 
50 °F
Flash Point(C) 
10 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

175188 Cyclopentene oxide, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?