EMAIL THIS PAGE TO A FRIEND
29930 Sigma-Aldrich

1-Cyclopropylethanol

purum, ≥97.0% (GC)

Synonym: α-Methylcyclopropanemethanol, Cyclopropyl methyl carbinol

  • CAS Number 765-42-4

  • Linear Formula C3H5CH(CH3)OH

  • Molecular Weight 86.13

  •  Beilstein Registry Number 1839704

  •  EC Number 212-145-9

  •  MDL number MFCD00001300

  •  PubChem Substance ID 329753399

Purchase

Properties

InChI Key   DKKVKJZXOBFLRY-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
impurities   ≤2% water
refractive index   n20/D 1.431(lit.)
  n20/D 1.431
bp   120-122 °C(lit.)
density   0.881 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1987 3/PG 3
WGK Germany 
3
Flash Point(F) 
99 °F
Flash Point(C) 
37 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

C119903 1-Cyclopropylethanol, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?