Synonym: TFA
CAS Number 76-05-1
Linear Formula CF3COOH
Molecular Weight 114.02
Beilstein Registry Number 742035
EC Number 200-929-3
MDL number MFCD00004169
PubChem Substance ID 24858046
Popular Documents: Specification Sheet (PDF)
| Related Categories | Amber Glass Bottles, Analytical Reagents, Analytical/Chromatography, Biotech, Biotech Solvents, |
| vapor density | 3.9 (vs air) |
| vapor pressure | 97.5 mmHg ( 20 °C) |
| type | for protein sequencing |
| assay | ≥99% |
| purified by | redistillation |
| impurities | <0.05% water |
| evapn. residue | <0.005% |
The effect of changing the direction of TFA delivery when sequencing proteins or peptides from PVDF membranes1.
25, 100, 500 g in glass bottle
50 mL in glass bottle
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?