EMAIL THIS PAGE TO A FRIEND
30017 Sigma-Aldrich

(+)-Cyclosativene

puriss., ≥99.0% (sum of enantiomers, GC)

  • CAS Number 22469-52-9

  • Empirical Formula (Hill Notation) C15H24

  • Molecular Weight 204.35

  •  Beilstein Registry Number 6791339

  •  MDL number MFCD00084734

Purchase

Properties

Related Categories Alkanes, Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Organic Building Blocks More...
InChI Key   XBWACJDEQIZTPR-YIZGKYRPSA-N
grade   puriss.
assay   ≥99.0% (sum of enantiomers, GC)
optical activity   [α]20/D +78±1°, neat
refractive index   n20/D 1.488(lit.)
  n20/D 1.488
bp   87 °C/10 mmHg(lit.)
density   0.929 g/mL at 25 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

448559 (+)-Cyclosativene, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?