• USA Home
  • 30130 - L-Cysteine hydrochloride monohydrate

EMAIL THIS PAGE TO A FRIEND
30130 Sigma-Aldrich

L-Cysteine hydrochloride monohydrate

≥99.0% (RT)

  • CAS Number 7048-04-6

  • Linear Formula HSCH2CH(NH2)COOH · HCl · H2O

  • Molecular Weight 175.63

  •  Beilstein Registry Number 5158059

  •  EC Number 200-157-7

  •  MDL number MFCD00065606

  •  PubChem Substance ID 329753440

Purchase

Properties

Related Categories Amino Acids, Amino Acids & Derivatives, Biochemicals and Reagents, Chemical Biology, Chemical Synthesis,
InChI Key   QIJRTFXNRTXDIP-JIZZDEOASA-N
assay   ≥99.0% (RT)
optical activity   [α]20/D +5.6±0.3°, c = 5% in 5 M HCl
impurities   ≤1% foreign amino acids
ign. residue   ≤0.1% (as SO4)
anion traces   sulfate (SO42-): ≤100 mg/kg
cation traces   Ca: ≤10 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cr: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  K: ≤50 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤5 mg/kg
  NH4+: ≤500 mg/kg
  Na: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg

Description

Biochem/physiol Actions

NMDA glutamatergic receptor antagonist.

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles

Articles

Dietary Antioxidants

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.1 During the oxi...
Biofiles Volume 2 Article 2
Keywords: Adhesion, Anti-inflammatory agents, Applications, Cancer, Cardiovascular, Catalysis, Diabetes, Diseases, Gene expression, Metabolism, Neurodegenerative Diseases, Peroxidations, Phosphorylations, Prebiotics, Probiotics, Reductions, Transcription, Transduction, Vitamins

Related Content

Amino Acids Reference Chart

The hydrophobicity index is a measure of the relative hydrophobicity, or how soluble an amino acid is in water. In a protein, hydrophobic amino acids are likely to be found in the interior, whereas h...

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C6852 L-Cysteine hydrochloride monohydrate, from non-animal source, cell culture tested, meets EP, USP testing specifications

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?