• USA Home
  • 30380 - Cytochalasin B from Drechslera dematioidea

EMAIL THIS PAGE TO A FRIEND
30380 Sigma-Aldrich

Cytochalasin B from Drechslera dematioidea

≥97.0% (HPLC)

Synonym: Phomin

  • CAS Number 14930-96-2

  • Empirical Formula (Hill Notation) C29H37NO5

  • Molecular Weight 479.61

  •  Beilstein Registry Number 1096207

  •  EC Number 239-000-2

  •  MDL number MFCD00077704

  •  PubChem Substance ID 329753487

Purchase

Properties

Related Categories Antibiotics, Antibiotics A to Z, Antibiotics A-F, Antibiotics by Application, Antineoplastic and Immunosuppressive Antibiotics,
InChI Key   GBOGMAARMMDZGR-TYHYBEHESA-N
assay   ≥97.0% (HPLC)
solubility   ethanol: 20 mg/mL clear, colorless
Mode of action   DNA synthesis | interferes
storage temp.   −20°C

Description

Biochem/physiol Actions

One of a group of fungal metabolites that interfere with a wide variety of cellular movements. Useful tool for characterizing some of the polymerization properties of actin, and in studies on cytokinesis. Probe for the two hexose-transport systems in rat L6 myoblasts.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1544PSN1 6.1 / PGII
WGK Germany 
3
RTECS 
RO0205000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?