EMAIL THIS PAGE TO A FRIEND
30405 Sigma-Aldrich

Daidzein

purum, ≥98.0% (TLC)

Synonym: 4′,7-Dihydroxy­iso­flavone, 7-Hydroxy-3-(4-hydroxy­phenyl)-4H-1-benzo­pyran-4-one, 7-Hydroxy-3-(4-hydroxy­phenyl)­chromone

  • CAS Number 486-66-8

  • Empirical Formula (Hill Notation) C15H10O4

  • Molecular Weight 254.24

  •  Beilstein Registry Number 231523

  •  EC Number 207-635-4

  •  MDL number MFCD00016954

  •  PubChem Substance ID 329753496

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Cell Biology, Chemical Class,
InChI Key   ZQSIJRDFPHDXIC-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (TLC)
storage temp.   2-8°C

Description

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Biochem/physiol Actions

Soy isoflavone daidzein protects against oxidative damage in liver cells induced by 7,12-dimethylbenz[a]anthracene (DMBA). Catalase and superoxide dismutase activity, down-regulated by DMBA, was restored by daidzein.

Price and Availability


All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3
RTECS 
DJ3100040

Documents

Certificate of Analysis

Protocols & Articles

Articles

Dietary Antioxidants

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.1 During the oxi...
Biofiles Volume 2 Article 2
Keywords: Adhesion, Anti-inflammatory agents, Applications, Cancer, Cardiovascular, Catalysis, Diabetes, Diseases, Gene expression, Metabolism, Neurodegenerative Diseases, Peroxidations, Phosphorylations, Prebiotics, Probiotics, Reductions, Transcription, Transduction, Vitamins

Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D7802 Daidzein, ≥98%, synthetic

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?