30410 Sigma-Aldrich

Sodium hypophosphite monohydrate

puriss. p.a., Reag. Ph. Eur., ≥99%

DOWNLOAD MSDS (PDF)
  • CAS Number 10039-56-2

  • Linear Formula NaH2PO2 · H2O

  • Molecular Weight 105.99

  •  EC Number 231-669-9

  •  MDL number MFCD06252379

  •  PubChem Substance ID 24858325

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Puriss p.a., Q-S, Puriss p.a. More...
grade   Reag. Ph. Eur.
  puriss. p.a.
assay   ≥99%
impurities   ≤0.001% ammonium (NH4)
pH   6-8 (20 °C, 5%)
anion traces   chloride (Cl-): ≤50 mg/kg
  phosphate, phosphite (as PO43-): ≤5000 mg/kg
  sulfate (SO42-): ≤100 mg/kg
cation traces   As: ≤1 mg/kg
  Ba: ≤50 mg/kg
  Ca: ≤200 mg/kg
  Cd: ≤5 mg/kg
  Co: ≤5 mg/kg
  Cu: ≤5 mg/kg
  Fe: ≤5 mg/kg
  Mg: ≤50 mg/kg
  Ni: ≤5 mg/kg
  Pb: ≤5 mg/kg
  Zn: ≤5 mg/kg

Price and Availability

Customers Also Viewed

Sodium hypophosphite

puriss., meets analytical specification of BPC63, anhydrous, 98-101%

Sodium hypophosphite monohydrate

puriss., meets analytical specification of DAC, 99-103%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?