Synonym: sec-Sodium phosphate, Disodium hydrogen phosphate dihydrate, Disodium phosphate, di-Sodium hydrogen phosphate dihydrate
CAS Number 10028-24-7
Linear Formula Na2HPO4 · 2H2O
Molecular Weight 177.99
EC Number 231-448-7
MDL number MFCD00149182
| Related Categories | Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Puriss p.a., Q-S, Puriss p.a. More... |
| grade | puriss. p.a. |
| assay | ≥99.5% |
| impurities | ≤0.001% ammonium (NH4) |
| ≤0.001% heavy metals (as Pb) | |
| pH | 9.0-9.2 (20 °C, 2%) |
| anion traces | chloride (Cl-): ≤10 mg/kg |
| sulfate (SO42-): ≤50 mg/kg |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?