30417 Sigma-Aldrich

Phosphoric acid

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%

DOWNLOAD MSDS (PDF)

Synonym: Orthophosphoric acid

  • CAS Number 7664-38-2

  • Linear Formula H3PO4

  • Molecular Weight 98.00

  •  Beilstein Registry Number 1921286

  •  EC Number 231-633-2

  •  MDL number MFCD00011340

  •  PubChem Substance ID 24858350

Properties

Related Categories ACS Grade, ACS Grade Acids & Bases, Acids & Bases, Analytical Reagents, Analytical Reagents for General Use,
grade   ACS reagent, reag. ISO, reag. Ph. Eur.
vapor density   3.4 (vs air)
vapor pressure   2.2 mmHg ( 20 °C)
  5 mmHg ( 25 °C)
grade   puriss. p.a.
assay   ≥85%
total impurities   reducing impurities, in accordance
  residual solvents, in accordance
  substances precipitated by ammonia, in accordance
  ≤0.001% KMnO4 red. matter (as O)
  ≤0.001% heavy metals (as Pb)
  ≤0.001% in water insoluble matters
  ≤0.001% volatile acids (as CH3COOH)
bp   158 °C(lit.)
mp   ~40 °C(lit.)
density   1.685 g/mL at 25 °C(lit.)
anion traces   chloride (Cl-): ≤2 mg/kg
  fluoride (F-): ≤1 mg/kg
  nitrate (NO3-): ≤3 mg/kg
  phosphite, hypophosphite (as H3PO3): ≤20 mg/kg
  phosphite, hypophosphite (as H3PO3): in accordance
  sulfate (SO42-): ≤30 mg/kg
cation traces   As: ≤0.5 mg/kg
  Ca: ≤10 mg/kg
  Cd: ≤1 mg/kg
  Co: ≤1 mg/kg
  Cr: ≤1 mg/kg
  Cu: ≤1 mg/kg
  Fe: ≤5 mg/kg
  K: ≤5 mg/kg
  Mg: ≤5 mg/kg
  Mn: ≤0.5 mg/kg
  Na: ≤20 mg/kg
  Ni: ≤1 mg/kg
  Pb: ≤1 mg/kg
  Sb: ≤2 mg/kg
  Zn: ≤2 mg/kg
suitability   passes test for appearance of solution
Gene Information   Research your gene in Your Favorite Gene powered by Ingenuity
human ... SRC(6714)

Description

Other Notes

Concentrated phosphoric acid is approx. 85% H3PO4 (aqueous).

Price and Availability

We Have the Silica You Need and a New Brochure

Silica Gel PurificationWhether you're looking for a specific brand, quality, or price, Sigma-Aldrich has the silica products to meet your unique purification or sample preparation requirements. Our new Silica Brochure makes it easy to find just the silica gel product you need.


Learn more.

Customers Also Purchased

32201Acetone

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10

CHROMASOLV®, for HPLC, ≥99.9%

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)

puriss. p.a., ACS reagent, reag. Ph. Eur., K ≤0.02%, ≥98%, pellets

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?