EMAIL THIS PAGE TO A FRIEND
30430 Sigma-Aldrich

Cytosine

≥99.0% (HPLC)

Synonym: 4-Amino-2-hydroxypyrimidine, 4-Aminopyrimidin-2-(1H)-one

  • CAS Number 71-30-7

  • Empirical Formula (Hill Notation) C4H5N3O

  • Molecular Weight 111.10

  •  Beilstein Registry Number 2637

  •  EC Number 200-749-5

  •  MDL number MFCD00006034

  •  PubChem Substance ID 24858360

Purchase

Properties

Related Categories Building Blocks, C4 to C5, Chemical Synthesis, Heterocyclic Building Blocks, Pyrimidines More...
InChI Key   OPTASPLRGRRNAP-UHFFFAOYSA-N
assay   ≥99.0% (HPLC)
mp   >300 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Quantity

λmax. 275 nm, log ε 10400 (in 0.1 N HCl)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
UW7350150

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

C3506 Cytosine, ≥99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?