• USA Home
  • 30518 - Decamethonium bromide

EMAIL THIS PAGE TO A FRIEND
30518 Sigma-Aldrich

Decamethonium bromide

for ion pair chromatography, ≥99.0% (AT)

Synonym: Decamethylene bis(trimethylammonium bromide), Decane-1,10-bis(trimethylammonium bromide)

  • CAS Number 541-22-0

  • Linear Formula (CH3)3N(Br)(CH2)10N(Br)(CH3)3

  • Molecular Weight 418.29

  •  Beilstein Registry Number 3728288

  •  EC Number 208-772-2

  •  MDL number MFCD00011779

  •  PubChem Substance ID 329753531

Purchase

Properties

Related Categories Acidic Ion Pair Reagents (Anionic), Analytical Reagents, Analytical/Chromatography, Chromatography Reagents & Solvents, Ion Pair Reagents More...
grade   for ion pair chromatography
InChI Key   HLXQFVXURMXRPU-UHFFFAOYSA-L
assay   ≥99.0% (AT)
mp   263-267 °C(lit.)
λ   10 % in H2O
UV absorption   λ: 240 nm Amax: 0.04
  λ: 250 nm Amax: 0.03
  λ: 260 nm Amax: 0.02
  λ: 500 nm Amax: 0.02
suitability   corresponds to standard for filter test

Description

General description

Decamethonium is a neuromuscular blocking agent.

Price and Availability


Laboratory Gloves

All labs need water
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
BP5950000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?