EMAIL THIS PAGE TO A FRIEND
30560 Sigma-Aldrich

Decane

purum, ≥95.0% (GC)

  • CAS Number 124-18-5

  • Linear Formula CH3(CH2)8CH3

  • Molecular Weight 142.28

  •  Beilstein Registry Number 1696981

  •  EC Number 204-686-4

  •  MDL number MFCD00008954

Purchase

Properties

vapor density   4.9 (vs air)
vapor pressure   1 mmHg ( 16.5 °C)
  3.77 mmHg ( 37.7 °C)
InChI Key   DIOQZVSQGTUSAI-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
autoignition temp.   410 °F
expl. lim.   2.6 %
refractive index   n20/D 1.411(lit.)
  n20/D 1.412
bp   174 °C(lit.)
mp   −30 °C(lit.)
density   0.73 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN 2247 3 / PGIII
WGK Germany 
1
RTECS 
HD6550000
Flash Point(F) 
114.8 °F
Flash Point(C) 
46 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?