EMAIL THIS PAGE TO A FRIEND
30640 Sigma-Aldrich

1-Decanethiol

technical, ≥95% (GC)

Synonym: DT, Decyl mercaptan, Mercaptan C10

  • CAS Number 143-10-2

  • Linear Formula CH3(CH2)9SH

  • Molecular Weight 174.35

  •  Beilstein Registry Number 1735223

  •  EC Number 205-584-2

  •  MDL number MFCD00004884

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Sulfur Compounds, Thiols/Mercaptans More...
InChI Key   VTXVGVNLYGSIAR-UHFFFAOYSA-N
grade   technical
assay   ≥95% (GC)
refractive index   n20/D 1.457(lit.)
  n20/D 1.458(lit.)
  n20/D 1.458
bp   114 °C/13 mmHg(lit.)
mp   −26 °C(lit.)
density   0.824 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3334
WGK Germany 
3
Flash Point(F) 
208.4 °F
Flash Point(C) 
98 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

D1602 1-Decanethiol, 96%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?