EMAIL THIS PAGE TO A FRIEND
30652 Sigma-Aldrich

1-Decene

purum, ≥95.0% (GC)

  • CAS Number 872-05-9

  • Linear Formula CH3(CH2)7CH=CH2

  • Molecular Weight 140.27

  •  Beilstein Registry Number 1737236

  •  EC Number 212-819-2

  •  MDL number MFCD00009577

  •  PubChem Substance ID 329753557

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
vapor density   4.84 (vs air)
vapor pressure   1.6 mmHg ( 23.8 °C)
InChI Key   AFFLGGQVNFXPEV-UHFFFAOYSA-N
grade   purum
assay   ≥95.0% (GC)
autoignition temp.   455 °F
refractive index   n20/D 1.421(lit.)
  n20/D 1.421
bp   166.5-173.5 °C(lit.)
mp   −66.3-−66 °C(lit.)
density   0.741 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Repeated exposure may cause skin dryness or cracking.
RIDADR 
UN 3295C 3 / PGIII
WGK Germany 
3
RTECS 
HE2071401
Flash Point(F) 
111.2 °F
Flash Point(C) 
44 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D1807 1-Decene, 94%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?