EMAIL THIS PAGE TO A FRIEND
30680 Sigma-Aldrich

Decylamine

purum, ≥98.0% (GC)

Synonym: 1-Aminodecane, NDA

  • CAS Number 2016-57-1

  • Linear Formula CH3(CH2)9NH2

  • Molecular Weight 157.30

  •  Beilstein Registry Number 1735220

  •  EC Number 217-957-7

  •  MDL number MFCD00008149

  •  PubChem Substance ID 24858500

Purchase

Properties

InChI Key   MHZGKXUYDGKKIU-UHFFFAOYSA-N
grade   purum
assay   ≥98.0% (GC)
refractive index   n20/D 1.436(lit.)
  n20/D 1.436
bp   216-218 °C(lit.)
mp   12-14 °C(lit.)
density   0.787 g/mL at 25 °C(lit.)

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN2922 - class 8 - PG 3 - EHS - acidic - Corrosive liquids, toxic, n.o.s., HI: all
WGK Germany 
3
RTECS 
HD6475000
Flash Point(F) 
192.2 °F
Flash Point(C) 
89 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

30692 Decylamine, ≥99.0% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?