30743 Sigma-Aldrich

Sulfuric acid

puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%

DOWNLOAD MSDS (PDF)
  • CAS Number 7664-93-9

  • Linear Formula H2SO4

  • Molecular Weight 98.08

  •  Beilstein Registry Number 2037554

  •  EC Number 231-639-5

  •  MDL number MFCD00064589

  •  PubChem Substance ID 24858593

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Puriss p.a., Q-S, Puriss p.a. More...
grade   ACS reagent, reag. ISO, reag. Ph. Eur.
vapor density   <0.3 (25 °C, vs air)
vapor pressure   1 mmHg ( 146 °C)
description   Nominally 95-98% H2SO4
grade   puriss. p.a.
assay   95.0-97.0%
impurities   ≤1 ppm heavy metals (as Pb)
  ≤2 ppm KMnO4 red. matter (as O)
  ≤2 ppm ammonium (NH4)
ign. residue   ≤5 ppm
bp   ~290 °C(lit.)
density   1.840 g/mL at 25 °C(lit.)
anion traces   chloride (Cl-): ≤0.1 ppm
  nitrate (NO3-): ≤0.2 ppm
  phosphate (PO43-): ≤0.5 mg/kg
cation traces   Ag: ≤0.02 mg/kg
  Al: ≤0.05 mg/kg
  As: ≤0.01 ppm
  Ba: ≤0.05 mg/kg
  Be: ≤0.01 mg/kg
  Bi: ≤0.05 mg/kg
  Ca: ≤0.2 mg/kg
  Cd: ≤0.02 mg/kg
  Co: ≤0.01 mg/kg
  Cr: ≤0.05 mg/kg
  Cu: ≤0.01 mg/kg
  Fe: ≤0.1 ppm
  Ge: ≤0.05 mg/kg
  Hg: ≤5 ppb
  K: ≤0.1 mg/kg
  Li: ≤0.01 mg/kg
  Mg: ≤0.05 mg/kg
  Mn: ≤0.01 mg/kg
  Mo: ≤0.02 mg/kg
  Na: ≤0.5 mg/kg
  Ni: ≤0.02 mg/kg
  Pb: ≤0.02 mg/kg
  Sr: ≤0.02 mg/kg
  Ti: ≤0.1 mg/kg
  Tl: ≤0.05 mg/kg
  V: ≤0.01 mg/kg
  Zn: ≤0.05 mg/kg
  Zr: ≤0.1 mg/kg

Description

Packaging

2.5, 4×2.5 L-GL in glass bottle

In PE-bottles;also available PE-bottles black (Material no. ending with PEBL; Glass-bottles (Material no. ending with *-GL), PE canister (Material no. ending with *-H)

Price and Availability

Customers Also Viewed

Hydrochloric acid

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10

Nitric acid

puriss. p.a., reag. ISO, reag. Ph. Eur., for determinations with dithizone, ≥65%

Sulfuric acid

ACS reagent, 95.0-98.0%

Sulfuric acid

ACS reagent, 95.0-98.0%

Sulfuric acid

puriss. p.a., 95-97% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?