307653 Sigma-Aldrich

Calcium hydrogenphosphate dihydrate

98%

DOWNLOAD MSDS (PDF)

Synonym: Calcium hydrogen phosphate dihydrate, Calcium phosphate dibasic dihydrate

  • CAS Number 7789-77-7

  • Linear Formula CaHPO4 · 2H2O

  • Molecular Weight 172.09

  •  EC Number 231-826-1

  •  MDL number MFCD00149621

  •  PubChem Substance ID 24858614

Properties

assay   98%
density   2.31 g/mL at 25 °C(lit.)

Description

Packaging

500 g in poly bottle

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Customers Also Viewed

Calcium phosphate

purum p.a., ≥96.0% (calc. as Ca3(PO4)2, KT)

Calcium phosphate dibasic

puriss. p.a., anhydrous, >98.5% (KT)

Calcium phosphate dibasic dihydrate

puriss., meets analytical specification of Ph. Eur., BP, USP, 98-102.5%

Dibasic Calcium Phosphate

meets USP testing specifications

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?