CAS Number 1309-48-4
Linear Formula MgO
Molecular Weight 40.30
EC Number 215-171-9
MDL number MFCD00011109
PubChem Substance ID 24858621
Popular Documents: Specification Sheet (PDF)
| Related Categories | Essential Chemicals, Inorganic Salts, Solutions and Reagents, Magnesium, Materials Science, Metal and Ceramic Science, |
| assay | 60-62% (RT) |
| mp | 2852 °C(lit.) |
| anion traces | sulfite and thiosulfate (as S2O32-: ≤1% |
| cation traces | Fe: ≤10 mg/kg |
25 g in glass bottle
500 g in poly bottle
Customers Also Viewed

puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 530, light, 98.0-100.5% (calc. for dried substance)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?