EMAIL THIS PAGE TO A FRIEND
309079 Sigma-Aldrich

Nitric acid

red, fuming, HNO3 >90 %

Purchase

Properties

Related Categories Acids, Acids & Bases, Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography,
vapor pressure   8 mmHg ( 20 °C)
composition   HNO3, >90%
impurities   5-10% dissolved oxides as N2O3
color   red
bp   120.5 °C(lit.)
density   1.48 g/mL at 20 °C(lit.)
storage temp.   2-8°C

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Packaging

100, 500 mL in PVC-coated bottle

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2032 6.1(5.1)(8)
WGK Germany 
2
RTECS 
QU5900000
Protocols & Articles

Related Content

Molarity Calculator and Normality Calculator for Acids and Bases

The molarity calculator tool provides lab-ready directions describing how to prepare an acid or base solution of specified Molarity (M) or Normality (N) from a concentrated acid or base solution.  To...

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?