EMAIL THIS PAGE TO A FRIEND
30925 Sigma-Aldrich

Cordycepin

≥98.0% (HPLC)

Synonym: 3′-Deoxyadenosine

  • CAS Number 73-03-0

  • Empirical Formula (Hill Notation) C10H13N5O3

  • Molecular Weight 251.24

  •  Beilstein Registry Number 35194

  •  EC Number 200-791-4

  •  MDL number MFCD00037998

  •  PubChem Substance ID 329753604

Purchase

Properties

Related Categories Antibiotics, Antibiotics A to Z, Antibiotics A-F, Antifungal, Biochemicals and Reagents,
InChI Key   OFEZSBMBBKLLBJ-BAJZRUMYSA-N
assay   ≥98.0% (HPLC)
mp   225-229 °C
Mode of action   DNA synthesis | interferes
  enzyme | inhibits
antibiotic activity spectrum   fungi

Description

Other Notes

Starting material for the synthesis of cordycepin triphosphate, an inhibitor of replicative synthesis

Biochem/physiol Actions

Cordycepin is an adenosine analogue that is readily converted to cordycepin 5′-triphosphate; can be used for 3′-end labeling of RNA.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
AU7358610

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

C3394 Cordycepin, from Cordyceps militaris

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?