• USA Home
  • 30926 - 2′-Deoxyadenosine 5′-monophosphate disodium salt

EMAIL THIS PAGE TO A FRIEND
30926 Sigma-Aldrich

2′-Deoxyadenosine 5′-monophosphate disodium salt

≥97.0% (HPLC)

Synonym: dAMP-Na2

  • CAS Number 2922-74-9

  • Empirical Formula (Hill Notation) C10H12N5Na2O6P

  • Molecular Weight 375.19

  •  Beilstein Registry Number 3583573

  •  EC Number 220-873-3

  •  MDL number MFCD00065990

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleosides, Nucleotides, Oligonucleotides, Nucleotides More...
InChI Key   SNYHHRDGEJPLGT-OJSHLMAWSA-L
assay   ≥97.0% (HPLC)
impurities   ≤20% water
solubility   H2O: soluble50 mg/mL, clear, colorless

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

D6375 2′-Deoxyadenosine 5′-monophosphate, Sigma Grade, 98-100%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?