EMAIL THIS PAGE TO A FRIEND
310352 Sigma-Aldrich

Iron

ACS reagent, granular, low in magnesium and manganese

Purchase

Properties

grade   ACS reagent
InChI Key   XEEYBQQBJWHFJM-UHFFFAOYSA-N
form   granular
resistivity   9.71 μΩ-cm
bp   2750 °C(lit.)
mp   1535 °C(lit.)
density   7.86 g/mL at 25 °C(lit.)
cation traces   Mg: ≤5 ppm
  Mn: ≤0.002%

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3089 4.1 / PGIII
WGK Germany 
nwg
RTECS 
NO4565500
Flash Point(F) 
69.8 °F
Flash Point(C) 
21 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

413054 Iron, granular, 10-40 mesh, >99.99% trace metals basis

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?