• USA Home
  • 31095 - 2′-Deoxyguanosine 5′-triphosphate trisodium salt solution

EMAIL THIS PAGE TO A FRIEND
31095 Sigma-Aldrich

2′-Deoxyguanosine 5′-triphosphate trisodium salt solution

≥95% (HPLC)

Synonym: dGTP-Na3

Purchase

Properties

InChI Key   IWGGLKOTEOCWQP-BIHLCPNHSA-K
assay   ≥95% (HPLC)
solubility   H2O: soluble50 mg/mL, clear, colorless
shipped in   dry ice
storage temp.   −20°C

Description

Other Notes

Substrate for DNA formation on model templates

Application

2′-Deoxyguanosine 5′-triphosphate trisodium salt is utilized for methods such as DNA sequencing, PCR and other DNA polymerase based DNA synthesis and repair techniques.

Biochem/physiol Actions

2′-Deoxyguanosine 5′-triphosphate is used in vivo synthesis for the synthesis of DNA via DNA polymerases.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?