31247 Sigma-Aldrich

Potassium chlorate

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%

DOWNLOAD MSDS (PDF)
  • CAS Number 3811-04-9

  • Linear Formula KClO3

  • Molecular Weight 122.55

  •  EC Number 223-289-7

  •  MDL number MFCD00011361

  •  PubChem Substance ID 24858894

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, O-P, Puriss p.a. ACS, Puriss p.a. ACS More...
grade   ACS reagent, reag. Ph. Eur.
  puriss. p.a.
assay   ≥99%
impurities   ≤0.0005% heavy metals (as Pb)
  ≤0.005% in water insoluble matters
mp   356 °C(lit.)
anion traces   bromate (BrO3-): ≤150 mg/kg
  chloride (Cl-): ≤10 mg/kg
  sulfate (SO42-): ≤20 mg/kg
cation traces   As: ≤0.5 mg/kg
  Ca: ≤20 mg/kg
  Fe: ≤3 mg/kg
  Mg: ≤10 mg/kg
  Na: ≤100 mg/kg

Price and Availability

Customers Also Viewed

Potassium chlorate

BioXtra, ≥99.0%

Potassium chlorate

ACS reagent, ≥99.0%

Potassium chlorate

puriss., 99-101%

Potassium chlorate

purum p.a., crystallized, ≥99.0% (T)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?