CAS Number 7447-40-7
Linear Formula KCl
Molecular Weight 74.55
EC Number 231-211-8
MDL number MFCD00011360
PubChem Substance ID 24858895
| Related Categories | Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, O-P, Puriss p.a., Puriss p.a. More... |
| grade | reag. ISO, reag. Ph. Eur. |
| puriss. p.a. | |
| assay | 99.5-100.5% |
| impurities | acidic or alkalic reac. substances, complies |
| ≤0.0005% heavy metals (as Pb) | |
| ≤0.001% total nitrogen (N) | |
| ≤0.003% free acid (as HCl) |
May be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins.
This product is also available in glass bottles.
Customers Also Viewed

puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?