EMAIL THIS PAGE TO A FRIEND
31340 Sigma-Aldrich

Chloroform-d

≥99.0 atom % D

Synonym: Deuterochloroform

  • CAS Number 865-49-6

  • Linear Formula CDCl3

  • Molecular Weight 120.38

  •  Beilstein Registry Number 1697633

  •  EC Number 212-742-4

Purchase

Properties

isotopic purity   ≥99.0 atom % D
contains   silver foil as stabilizer
impurities   ≤0.02% HDO (NMR)
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 1888 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?