EMAIL THIS PAGE TO A FRIEND
31399 Sigma-Aldrich

Dextrangel N 25

for column chromatography

Purchase

Properties

Related Categories Analytical/Chromatography, Gel Filtration, LPLC Separation Media, LPLC/TLC/Paper Chromatography, Sephadex More...
grade   for column chromatography

Description

General description

Crosslinked dextran for gel filtration with a grain size (dry) of 40-150 μm and an exclusion limit of Mr~6000 (dextrans, proteins);
useful for desalting and group separations
Water uptake: ~2.5 ml/g; gel bed volume ~5 ml/g
Hydrodynamic properties:
270 ml·h-1·cm-2 at 1 bar pressure (column size 50×8 cm)

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?