EMAIL THIS PAGE TO A FRIEND
31409 Sigma-Aldrich

Magnesia grooves

  • Empirical Formula (Hill Notation) MgO

  • Molecular Weight 40.30

  •  EC Number 215-171-9

  •  MDL number MFCD00011109

Purchase

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Packaging

package of 50 pieces

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
OM3850000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?