EMAIL THIS PAGE TO A FRIEND
31433 Sigma-Aldrich

DEV Gelatin Agar

for microbiology

Purchase

Properties

Related Categories A - M, All Solid Media/Agars, Bacteria (General Media), Biochemical Identification Media by Test, Biochemical Identification Media, A to Z,
grade   for microbiology
shelf life   limited shelf life, expiry date on the label
composition   agar, 15.0 g/L
  gelatin, 10.0 g/L
  meat extract, 10.0 g/L
  meat peptone, 10.0 g/L
  sodium chloride, 5.0 g/L
final pH   7.3±0.2 (25 °C)
suitability   Proteolytic Bacteria
  nonselective and differential for Bacteria (General Media)

Description

Application

Suitable agar for enumeration, including gelatine utilizing microorganisms, in water acc. to the water regulations of the "Deutschen Einheitsverfahren".

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

EZ-Fit™ Filtration Products for Bioburden Testing

The EZ family of products has been designed to optimize and streamline the bioburden workflow. The family includes:
Keywords: Filtration

Microbiology Introduction

An initial aim of all microbiologists is the reproducible growth of their microbial cultures, no matter whether the microorganisms are of natural origin or have been genetically engineered by man. Re...
Keywords: Absorption, Aerobic, Anaerobic, Antibiotics, Antimicrobials, Buffers, Culture media, Diffusion, Environmental, Filtration, Genetic, Growth factors, Metabolism, Phase transitions, Precipitation, Reductions, Respiration, Sample preparations, Vitamins

Microbiology Theory: Media Preparation

Microorganisms need nutrients, a source of energy and certain environmental conditions in order to grow and reproduce. In the environment, microbes have adapted to the habitats most suitable for thei...
Keywords: Culture media, Digestions, Environmental, Filtration, Growth factors, Indicators, Microbiology, Phase transitions, Precipitation, Vitamins

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?