31437 Sigma-Aldrich

Sodium bicarbonate

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.7%, powder

DOWNLOAD MSDS (PDF)

Synonym: Sodium hydrogen carbonate

  • CAS Number 144-55-8

  • Linear Formula NaHCO3

  • Molecular Weight 84.01

  •  Beilstein Registry Number 4153970

  •  EC Number 205-633-8

  •  MDL number MFCD00003528

  •  PubChem Substance ID 24858955

Properties

Related Categories Analytical Reagents, Analytical Reagents for General Use, Analytical/Chromatography, Puriss p.a. ACS, Q-S, Puriss p.a. ACS More...
grade   ACS reagent, reag. Ph. Eur.
  puriss. p.a.
assay   ≥99.7%
form   powder
impurities   residual solvents, complies
  ≤0.0005% ammonium (NH4)
  ≤0.003% S-compounds (as SO4)
  ≤0.005% I red. matters (as SO2)
  ≤0.015% in water insoluble matters
pH   ≤8.6 (20 °C, 5%)
pKa (25 °C)   (1) 6.37, (2) 10.25 (carbonic acid)
solubility   H2O: soluble0.5 g/10 mL, clear, colorless
anion traces   chloride (Cl-): ≤10 mg/kg
  phosphate (PO43-): ≤10 mg/kg
cation traces   As: 2 ppm (MHS-AAS)
  Ca: ≤100 ppm
  Cd: ≤5 ppm
  Co: ≤50 ppm
  Cu: ≤5 ppm
  Fe: ≤5 ppm
  K: ≤50 ppm
  Mg: ≤50 ppm
  Ni: ≤50 ppm
  Pb: ≤5 ppm
  Zn: ≤5 ppm
suitability   complies for appearance of solution

Price and Availability

Customers Also Viewed

for microbiology

Sodium bicarbonate

powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture

Sodium bicarbonate

ACS reagent, 99.7-100.3%

Sodium bicarbonate

puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder

Sodium carbonate

puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., anhydrous, ≥99.8% (calc. to the dried substance)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?