• USA Home
  • 31444 - Sodium nitroprusside dihydrate

EMAIL THIS PAGE TO A FRIEND
31444 Sigma-Aldrich

Sodium nitroprusside dihydrate

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%

Synonym: Nitroprusside sodium, SNP, Sodium nitroferricyanide, Sodium pentacyanonitrosylferrate

Purchase

Properties

grade   ACS reagent, reag. Ph. Eur.
InChI Key   OIRZWVYIQXBRFC-UHFFFAOYSA-N
grade   puriss. p.a.
assay   ≥99%
impurities   ≤0.01% hexacyanoferrate(III) ([Fe(CN)6]3-)
  ≤0.02% hexacyanoferrate(II) ([Fe(CN)6]4-)
anion traces   chloride (Cl-): ≤100 mg/kg
  sulfate (SO42-): ≤100 mg/kg

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Application

Sodium nitroprusside dihydrate may be used in the conditioned medium for in vitro activation assay.

General description

Sodium nitroprusside dihydrate (disodium pentacyanonitrosylferrate(2-)dihydrate, SNP) molecule contains a ferrous ion in the center, which is surrounded by five cyanide groups and a nitrosyl group. Sodium nitroprusside has clinical applications and acts as a rapid-acting vasodilator. It is also widely employed as nitric oxide (NO) donor in various experimental models. Two metastable states of sodium nitroprusside dihydrate, Na2[Fe(CN)5NO].2H2O have been identified and their geometries have been studied by X-ray diffraction.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3288 6.1 / PGIII
WGK Germany 
3
RTECS 
LJ8925000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?